Calcium ≥99.99%, crystalline dendritic solid

Supplier: BeanTown Chemical


Ampuled under argon

130585-25G 130585-5G
BT130585-25GEA 825 USD
BT130585-25G BT130585-5G
Calcium ≥99.99%, crystalline dendritic solid
Calcium

CAS: 7440-70-2; EC No: 213-179-5; MDL No: MFCD00085314; RTECS: EV8040000 UN No: UN1401; Haz Class: 4.3; Packing Group: II Solid; Molecular Formula: Ca; MW: 40.08 Melting Point: 850°; Boiling Point: 1484° Density (g/mL): 1.54


  • RTECS: EV8040000

Formula: Ca
MW: 40.08 g/mol
Boiling Pt: 1484
Melting Pt: 850
Density: 1.54 g/mL
MDL Number: MFCD00085314
CAS Number: 7440-70-2
UN: 1401
ADR: 4.3 II
Merck Index: 13,01644

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR