Lanthanum(III) nitrate hexahydrate ≥99.9%
Supplier: BeanTown Chemical
130470-100G
130470-500G
BT130470-100GEA
71.86
USD
BT130470-100G
BT130470-500G
Lanthanum(III) nitrate hexahydrate ≥99.9%
Lanthanum(III) nitrate hexahydrate
CAS: 10277-43-7; EC No: 233-238-0; MDL No: MFCD00149751; RTECS: OE5075000
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: La(NO3)3·6H2O; MW: 433.01
Melting Point: 65-68°
- RTECS: OE5075000
- Crystalline
Formula:
La(NO₃)₃·6H₂O MW: 433.01 g/mol Boiling Pt: 126 Melting Pt: 65…68 °C |
MDL Number:
MFCD00149751 CAS Number: 10277-43-7 EINECS: 233-238-0 UN: 1477 ADR: 5.1 III Merck Index: 14,05363 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...