Propylamine ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Propylamine, 1-Aminopropane

130385-1L 130385-250ML
BT130385-1LEA 95 USD
BT130385-1L BT130385-250ML
Propylamine ≥99%
Propylamine

CAS: 107-10-8; EC No: 203-462-3; MDL No: MFCD00008205; RTECS: UH9100000 UN No: UN1277; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: CH3CH2CH2NH2; Molecular Formula: C3H9N; MW: 59.11 Melting Point: -83°; Boiling Point: 48°; Flash point: -30°C (-22°F) Density (g/mL): 0.719; Refractive Index: 1.388 Air Sensitive


  • RTECS: UH9100000
  • Refractive index: 1.388
  • Air Sensitive
  • Liquid

Formula: H₃CCH₂CH₂NH₂
MW: 59.11 g/mol
Boiling Pt: 48 °C
Melting Pt: –83 °C
Density: 0.719 g/mL
Flash Pt: –30 °C (–22 °F)
MDL Number: MFCD00008205
CAS Number: 107-10-8
EINECS: 203-462-3
UN: 1277
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR