Pyruvic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid
130360-100G
130360-2.5KG
130360-500G
BT130360-100GEA
71.86
USD
BT130360-100G
BT130360-2.5KG
BT130360-500G
Pyruvic acid ≥98%
Pyruvic acid
CAS: 127-17-3; EC No: 204-824-3; MDL No: MFCD00002585; RTECS: UZ0829800
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: CH3COCOOH; Molecular Formula: C3H4O3; MW: 88.06
Melting Point: 11-12°; Boiling Point: 164-166°; Flash point: 82°C (180°F)
Density (g/mL): 1.267; Refractive Index: 1.428
- RTECS: UZ0829800
- Refractive index: 1.428
- Liquid
Formula:
C₃H₄O₃ MW: 88.06 g/mol Boiling Pt: 164…166 °C Melting Pt: 11…12 °C Density: 1.267 g/mL Flash Pt: 82 °C (180 °F) |
MDL Number:
MFCD00002585 CAS Number: 127-17-3 EINECS: 204-824-3 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...