Butyl acetate ≥99%

Supplier: BeanTown Chemical

Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

130235-2.5L 130235-500ML
BT130235-2.5LEA 128.75 USD
BT130235-2.5L BT130235-500ML
Butyl acetate ≥99%
Butyl acetate

CAS: 123-86-4; EC No: 204-658-1; MDL No: MFCD00009445; RTECS: AF7350000 UN No: UN1123; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3COO(CH2)3CH3; Molecular Formula: C6H12O2; MW: 116.16 Melting Point: -78°; Boiling Point: 124-126°; Flash point: 23°C (73°F) Density (g/mL): 0.881; Refractive Index: 1.394


  • RTECS: AF7350000
  • Refractive index: 1.394
  • Liquid

Formula: CH₃COO(CH₂)₃CH₃
MW: 116.16 g/mol
Boiling Pt: 124…126 °C
Melting Pt: –78 °C
Density: 0.881 g/mL
Flash Pt: 23 °C (73 °F)
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR