Anisole ≥99%

Supplier: BeanTown Chemical

Synonyms: Methyl phenyl ether, Methoxybenzene

130230-2.5KG 130230-500G
BT130230-2.5KGEA 163.11 USD
BT130230-2.5KG BT130230-500G
Anisole ≥99%
Anisole

CAS: 100-66-3; EC No: 202-876-1; MDL No: MFCD00008354; RTECS: BZ8050000 UN No: UN2222; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H8O; MW: 108.14 Melting Point: -37°; Boiling Point: 154°; Flash point: 51°C (124°F) Density (g/mL): 0.995; Refractive Index: 1.516


  • RTECS: BZ8050000
  • Refractive index: 1.516
  • Liquid

Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 154 °C
Melting Pt: –37 °C
Density: 0.995 g/mL
Flash Pt: 51 °C (124 °F)
MDL Number: MFCD00008354
CAS Number: 100-66-3
EINECS: 202-876-1
UN: 2222
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d32206d4bb9dbd12360d11d39b31237d54a8dcf8&vid=286a07a5e34a1ec55ff6339785af95d9a4326825&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR