Tin(II) telluride ≥99.999%

Supplier: BeanTown Chemical

130190-10G 130190-50G
BT130190-10GEA 88 USD
BT130190-10G BT130190-50G
Tin(II) telluride ≥99.999%
Tin(II) telluride

CAS: 12040-02-7; EC No: 234-914-8; MDL No: MFCD00049543 UN No: UN3288; Haz Class: 6.1; Packing Group: III 3-12mm Pieces; Molecular Formula: SnTe; MW: 246.31 Melting Point: 780° Density (g/mL): 6.48


  • 3-12mm Pieces

Formula: SnTe
MW: 246.31 g/mol
Melting Pt: 790
Density: 6.48 g/mL
MDL Number: MFCD00049543
CAS Number: 12040-02-7
EINECS: 234-914-8
UN: 3288
ADR: 6.1 III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR