Ethylene glycol monovinyl ether ≥97%
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Vinyl Cellosolve, Ethylene glycol vinyl ether, 2-Vinyloxyethanol, Vinyl Glycol, 2-(Vinyloxy)ethan-1-ol
130185-10ML
130185-50ML
BT130185-10MLEA
20
USD
BT130185-10ML
BT130185-50ML
Ethylene glycol monovinyl ether ≥97%
Ethylene glycol monovinyl ether
CAS: 764-48-7; EC No: 212-124-4; MDL No: MFCD00192151; RTECS: KM5495000
UN No: UN3271; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: H2C=CHOCH2CH2OH; Molecular Formula: C4H8O2; MW: 88.11
Boiling Point: 143°; Flash point: 49°C (120°F)
Density (g/mL): 0.982; Refractive Index: 1.436
- Refractive index: 1.436
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 143 °C Density: 0.982 g/mL Flash Pt: 49 °C (120 °F) |
MDL Number:
MFCD00192151 CAS Number: 764-48-7 UN: 3271 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...