tert-Butyl hydroperoxide ≥70% (w/w) in water
Supplier: BeanTown Chemical
|
Danger
|
CAS: 75-91-2; EC No: 200-915-7; MDL No: MFCD00002130; RTECS: EQ4900000
UN No: UN3109; Haz Class: 5.2 (8); Packing Group: II
Liquid; Linear Formula: (CH3)3COOH; Molecular Formula: C4H10O2; MW: 90.12
Melting Point: -3°; Boiling Point: 96°; Flash point: 42°C (108°F)
Density (g/mL): 0.937; Refractive Index: 1.386
- RTECS: EQ4900000
- Refractive index: 1.386
- Liquid
Formula:
C₄H₁₀O₂ Boiling Pt: 96 °C Melting Pt: –3 °C Density: 0.937 g/mL Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002130 CAS Number: 75-91-2 EINECS: 200-915-7 UN: 3109 ADR: 5.2 (8), |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...