Copper(I) iodide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Cuprous iodide
129985-1KG
129985-250G
BT129985-1KGEA
271
USD
BT129985-1KG
BT129985-250G
Copper(I) iodide ≥98%
Copper(I) iodide
CAS: 7681-65-4; EC No: 231-674-6; MDL No: MFCD00010978
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: CuI; MW: 190.45
Melting Point: 605°
Density (g/mL): 5.62
Air Sensitive, Light Sensitive, Moisture Sensitive
- Air Sensitive, Light Sensitive, Moisture Sensitive
- Powder
Formula:
CuI MW: 190.45 g/mol Melting Pt: 605 °C Density: 5.62 g/mL |
MDL Number:
MFCD00010978 CAS Number: 7681-65-4 EINECS: 231-674-6 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...