Samarium ≥99.9% (trace rare earth metals basis), Powder -40 mesh

Supplier: BeanTown Chemical


ampouled under argon

129960-10G 129960-50G
BT129960-10GEA 169 USD
BT129960-10G BT129960-50G
Samarium ≥99.9% (trace rare earth metals basis), Powder -40 mesh
Samarium

CAS: 7440-19-9; EC No: 231-128-7; MDL No: MFCD00011233 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: Sm; MW: 150.36 Melting Point: 1074°; Boiling Point: 1794° Density (g/mL): 7.47


  • Powder

Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1794 °C
Melting Pt: 1074 °C
Density: 7.47 g/mL
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR