Dinickel boride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Nickel boride
129940-100G
129940-25G
BT129940-100GEA
189
USD
BT129940-100G
BT129940-25G
Dinickel boride ≥99%
Dinickel boride
CAS: 12007-01-1; EC No: 234-494-6; MDL No: MFCD00084757
UN No: UN3077; Haz Class: 9; Packing Group: III
-325 Mesh Granular; Linear Formula: Ni2B; MW: 128.23
- -325 Mesh Granular
Formula:
BNi₂ MW: 128.23 g/mol |
MDL Number:
MFCD00084757 CAS Number: 12007-01-1 EINECS: 234-494-6 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...