Silver fluoride ≥98%

Supplier: BeanTown Chemical

Synonyms: Silver monofluoride, Silver(I) fluoride, Argentous fluoride

129915-25G 129915-5G
BT129915-25GEA 217 USD
BT129915-25G BT129915-5G
Silver fluoride ≥98%
Silver fluoride

CAS: 7775-41-9; EC No: 231-895-8; MDL No: MFCD00003410; RTECS: VW4250000 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: AgF; MW: 126.87 Melting Point: 435°; Boiling Point: 1150° Density (g/mL): 5.852 Hygroscopic , Light Sensitive


  • RTECS: VW4250000
  • Hygroscopic, Light Sensitive
  • Crystalline

Formula: AgF
MW: 126.87 g/mol
Boiling Pt: 1150 °C
Melting Pt: 435 °C
Density: 5.852 g/mL
MDL Number: MFCD00003410
CAS Number: 7775-41-9
EINECS: 231-895-8
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR