Propargyl amine ≥98%
Supplier: BeanTown Chemical
CAS: 2450-71-7; EC No: 219-513-8; MDL No: MFCD00008198; RTECS: UK5250000
UN No: UN3286; Haz Class: 3 (6.1,8); Packing Group: II
Liquid; Linear Formula: HC≡CCH2NH2; Molecular Formula: C3H5N; MW: 55.08
Boiling Point: 83°; Flash point: 6°C (43°F)
Density (g/mL): 0.86; Refractive Index: 1.449
Hygroscopic , Light Sensitive
- RTECS: UK5250000
- Refractive index: 1.449
- Hygroscopic, Light Sensitive
- Liquid
Formula:
C₃H₅N Boiling Pt: 83 °C Density: 0.86 g/mL Flash Pt: 6 °C (43 °F) |
MDL Number:
MFCD00008198 CAS Number: 2450-71-7 EINECS: 219-513-8 UN: 3286 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...