Dibutyltin dilaurate ≥95%
Supplier: BeanTown Chemical
CAS: 77-58-7; EC No: 201-039-8; MDL No: MFCD00008963; RTECS: WH7000000
UN No: UN2788; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C32H64O4Sn; MW: 631.55
Boiling Point: 204°/12 mmHg; Flash point:
- RTECS: WH7000000
- Refractive index: 1.471
- Liquid
Formula:
C₃₂H₆₄O₄Sn Boiling Pt: 204 °C/12 mmHg Density: 1.066 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00008963 CAS Number: 77-58-7 EINECS: 201-039-8 UN: 2788 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...