4-Pentynoic acid ≥98%
Supplier: BeanTown Chemical
CAS: 6089-09-4; EC No: 228-028-0; MDL No: MFCD00004407; RTECS: SC4751000
UN No: UN3261; Haz Class: 8; Packing Group: II
Powder/Flakes; Molecular Formula: C5H6O2; MW: 98.10
Melting Point: 54-57°; Boiling Point: 110°/30 mmHg; Flash point: 75°C (167°F)
Air Sensitive, Hygroscopic, Light Sensitive
- RTECS: SC4751000
- Air Sensitive, Hygroscopic, Light Sensitive
- Powder/Flakes
Formula:
C₅H₆O₂ Boiling Pt: 110 °C/30 mmHg Melting Pt: 54…57 °C Flash Pt: 75 °C (167 °F) |
MDL Number:
MFCD00004407 CAS Number: 6089-09-4 EINECS: 228-028-0 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...