4-Chloro-2-(methylthio)pyrimidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Chloro-2-methylthiopyrimidine

129710-10G 129710-250G 129710-50G
BT129710-10GEA 61 USD
BT129710-10G BT129710-250G BT129710-50G
4-Chloro-2-(methylthio)pyrimidine ≥98%
4-Chloro-2-(methylthio)pyrimidine

CAS: 49844-90-8; EC No: 256-500-6; MDL No: MFCD00006083 UN No: UN1760; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C5H5ClN2S; MW: 160.62 Melting Point: -2°; Boiling Point: 139-140°/36 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.381; Refractive Index: 1.6 Moisture Sensitive


  • Refractive index: 1.6
  • Moisture Sensitive
  • Liquid

Formula: C₅H₅ClN₂S
MW: 160.63 g/mol
Boiling Pt: 139…140 °C/36 mmHg
Melting Pt: –2 °C
Density: 1.381 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00006083
CAS Number: 49844-90-8
EINECS: 256-500-6
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR