1,3-Dithiane ≥98%

Supplier: BeanTown Chemical

129685-100G 129685-25G 129685-5G
BT129685-100GEA 553 USD
BT129685-100G BT129685-25G BT129685-5G
1,3-Dithiane ≥98%
1,3-Dithiane

CAS: 505-23-7; EC No: 208-006-7; MDL No: MFCD00006654; RTECS: JO5070000 UN No: UN3335; Haz Class: 9; Packing Group: III Solid; Molecular Formula: C4H8S2; MW: 120.24 Melting Point: 52-54°; Flash point: 90°C (194°F)


  • RTECS: JO5070000
  • Solid

Formula: C₄H₈S₂
MW: 120.24 g/mol
Melting Pt: 52…54 °C
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00006654
CAS Number: 505-23-7
EINECS: 208-006-7
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR