Dibutyltin diacetate ≥95%

Supplier: BeanTown Chemical

129670-1KG 129670-250G 129670-50G
BT129670-1KGEA 268 USD
BT129670-1KG BT129670-250G BT129670-50G
Dibutyltin diacetate ≥95%
Dibutyltin diacetate

CAS: 1067-33-0; EC No: 213-928-8; MDL No: MFCD00008697 UN No: UN2788; Haz Class: 6.1; Packing Group: II Liquid; Linear Formula: (CH3CH2CH2CH2)2Sn(OCOCH3)2; Molecular Formula: C12H14O4Sn; MW: 351.01 Melting Point: 7-10°; Boiling Point: 142-145°/10 mmHg; Flash point: 105°C (221°F) Density (g/mL): 1.32; Refractive Index: 1.471


  • Refractive index: 1.471
  • Liquid

Formula: C₁₂H₂₄O₄Sn
Boiling Pt: 142…145 °C/10 mmHg
Melting Pt: 7…10 °C
Density: 1.32 g/mL
Flash Pt: 105 °C (221 °F)
MDL Number: MFCD00008697
CAS Number: 1067-33-0
EINECS: 213-928-8
UN: 2788
ADR: 6.1,II

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR