Nitric acid 68 - 70%, Environmental Grade
Supplier: BeanTown Chemical
CAS: 7697-37-2; EC No: 231-714-2; MDL No: MFCD00011349; RTECS: QU5775000
UN No: UN2031; Haz Class: 8 (5.1); Packing Group: II
Liquid, double sub-boiling quartz distillation; Molecular Formula: HNO3 ; MW: 63.01
Boiling Point: 120.5°
Density (g/mL): 1.413
- RTECS: QU5775000
- Liquid, double sub-boiling quartz distillation
Formula:
HNO₃ Boiling Pt: 120.5 °C Density: 1.413 g/mL |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...