Titanium carbide, nanopowder

Supplier: BeanTown Chemical

129635-100G 129635-25G 129635-5G
BT129635-100GEA 524 USD
BT129635-100G BT129635-25G BT129635-5G
Titanium carbide, nanopowder
Titanium carbide

CAS: 12070-08-5; EC No: 235-120-4; MDL No: MFCD00011268; RTECS: XR1903500 UN No: UN3178; Haz Class: 4.1; Packing Group: III 30-50nm APS Powder, S.A. 35-45 m2/g; Linear Formula: TIC; MW: 59.89 Melting Point: 3140°; Boiling Point: 4820° Density (g/mL): 4.93


  • RTECS: XR1903500
  • 30-50nm APS Powder, S.A. 35-45 m²/g

Formula: CTi
MW: 59.89 g/mol
Boiling Pt: 4820 °C
Melting Pt: 3140 °C
Density: 4.93 g/mL
MDL Number: MFCD00011268
CAS Number: 12070-08-5
EINECS: 235-120-4
UN: 3178
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR