Vanadium(V) oxide ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Vanadium pentoxide, di-Vanadium pentaoxide
129625-1G
129625-25G
129625-5G
BT129625-1GEA
25
USD
BT129625-1G
BT129625-25G
BT129625-5G
Vanadium(V) oxide ≥99.99% (trace metals basis)
Vanadium(V) oxide
CAS: 1314-62-1; EC No: 215-239-8; MDL No: MFCD00011457; RTECS: YW2125000
UN No: UN2862; Haz Class: 6.1; Packing Group: III
-22 Mesh Powder; Linear Formula: V2O5; MW: 181.88
Melting Point: 690°; Boiling Point: 1750° (decomposes)
Density (g/mL): 3.357
- RTECS: YW2125000
- -22 Mesh Powder
Formula:
O₅V₂ MW: 181.88 g/mol Boiling Pt: 1750 °C (dec.) Melting Pt: 690 °C Density: 3.357 g/mL |
MDL Number:
MFCD00011457 CAS Number: 1314-62-1 EINECS: 215-239-8 UN: 2862 ADR: 6.1,III |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...