Selenium dioxide ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
CAS: 7446-08-4; EC No: 231-194-7; MDL No: MFCD00003562
UN No: UN3283; Haz Class: 6.1; Packing Group: II
Powder/Lump; Linear Formula: SeO2; MW: 110.96
Melting Point: 315° (sublimes)
Density (g/mL): 3.95
Hygroscopic
- Hygroscopic
- Powder/Lump
Formula:
SeO₂ Melting Pt: 315 °C (subl.) Density: 3.95 g/mL |
MDL Number:
MFCD00003562 CAS Number: 7446-08-4 EINECS: 231-194-7 UN: 3283 ADR: 6.1,II |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...