2-Fluoro-5-nitrotoluene ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Fluoro-2-methyl-4-nitrobenzene

129540-10G 129540-50G
BT129540-10GEA 60 USD
BT129540-10G BT129540-50G
2-Fluoro-5-nitrotoluene ≥98%
2-Fluoro-5-nitrotoluene

CAS: 455-88-9; EC No: 207-251-7; MDL No: MFCD00007284 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C7H6FNO2; MW: 155.13 Melting Point: 38-40°; Boiling Point: 99-100°/13 mmHg; Flash point: 105°C (221°F)


  • Crystalline

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 99…100 °C/13 mmHg
Melting Pt: 38…40 °C
Flash Pt: 105 °C (221 °F)
MDL Number: MFCD00007284
CAS Number: 455-88-9
EINECS: 207-251-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR