Sodium nitrite ≥97.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Diazotizing salts
129535-100G
129535-25G
129535-500G
BT129535-100GEA
39.16
USD
BT129535-100G
BT129535-25G
BT129535-500G
Sodium nitrite ≥97.0% ACS
Sodium nitrite
CAS: 7632-00-0; EC No: 231-555-9; MDL No: MFCD00011118; RTECS: RA1225000
UN No: UN1500; Haz Class: 5.1 (6.1); Packing Group: III
Solid; Linear Formula: NaNO2; MW: 69.00
Melting Point: 271°; Boiling Point: 320°
Density (g/mL): 2.168
Hygroscopic
- RTECS: RA1225000
- Hygroscopic
- Solid
Formula:
NaNO₂ MW: 69 g/mol Boiling Pt: 320 °C Melting Pt: 271 °C Density: 2.168 g/mL |
MDL Number:
MFCD00011118 CAS Number: 7632-00-0 EINECS: 231-555-9 UN: 1500 ADR: 5.1,III |
Specification Test Results
Assay | 97.0% min. |
Insoluble matter | ≤0.01% |
Chloride (Cl) | ≤0.005% |
Sulfate (SO4) | ≤0.01% |
Calcium (Ca) | ≤0.01% |
Potassium (K) | ≤0.005% |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤0.001% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...