Quinhydrone ≥97%
Supplier: BeanTown Chemical
Synonyms:
Benzoquinone-hydroquinone, p-Benzoquinone hydroquinone complex, 2,5-Cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol (1:1)
129530-100G
129530-25G
BT129530-100GEA
97.36
USD
BT129530-100G
BT129530-25G
Quinhydrone ≥97%
Quinhydrone
CAS: 106-34-3; EC No: 203-387-6; MDL No: MFCD00010310; RTECS: VA4550000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C12H10O4; MW: 218.21
Melting Point: 167-172°
Density (g/mL): 1.4
Air Sensitive, Light Sensitive
- RTECS: VA4550000
- Air Sensitive, Light Sensitive
- Crystalline
Formula:
C₆H₄(OH)₂·C₆H₄O₂ MW: 218.21 g/mol Melting Pt: 167…172 °C Density: 1.4 g/mL |
MDL Number:
MFCD00010310 CAS Number: 106-34-3 EINECS: 203-387-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...