3,5-Dichloro-4-hydroxybenzonitrile ≥97%

Supplier: BeanTown Chemical

Synonyms: Chloroxynil

129525-1G 129525-5G
BT129525-1GEA 169 USD
BT129525-1G BT129525-5G
3,5-Dichloro-4-hydroxybenzonitrile ≥97%
3,5-Dichloro-4-hydroxybenzonitrile

CAS: 1891-95-8; EC No: 217-572-4; MDL No: MFCD00002177 UN No: UN3439; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C7H3Cl2NO; MW: 188.01 Melting Point: 138-142°


  • Powder

Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
Melting Pt: 138…142 °C
MDL Number: MFCD00002177
CAS Number: 1891-95-8
EINECS: 217-572-4
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR