Selenium disulfide ≥97%
Supplier: BeanTown Chemical
129440-250G
129440-50G
129440-5G
BT129440-250GEA
412
USD
BT129440-250G
BT129440-50G
BT129440-5G
Selenium disulfide ≥97%
Selenium disulphide
CAS: 7488-56-4; EC No: 231-303-8; MDL No: MFCD00011227
UN No: UN2657; Haz Class: 6.1; Packing Group: II
Powder; Linear Formula: SeS2; MW: 143.09
Melting Point:
- Powder
Formula:
S₂Se Melting Pt: <100 °C |
MDL Number:
MFCD00011227 CAS Number: 7488-56-4 EINECS: 231-303-8 UN: 2657 ADR: 6.1,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...