(±)-sec-Butylamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
(±)-2-Butylamine, (±)-2-Aminobutane
129425-2.5L
129425-500ML
BT129425-2.5LEA
343
USD
BT129425-2.5L
BT129425-500ML
(±)-sec-Butylamine ≥98%
(±)-sec-Butylamine
CAS: 13952-84-6; EC No: 237-732-7; MDL No: MFCD00008094; RTECS: EO3325000
UN No: UN2733; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: CH3CH2CH(NH2)CH3; Molecular Formula: C4H11N; MW: 73.14
Melting Point: -72°; Boiling Point: 63°; Flash point: -10°C (2°F)
Density (g/mL): 0.724; Refractive Index: 1.393
Air Sensitive
- RTECS: EO3325000
- Refractive index: 1.393
- Air Sensitive
- Liquid
Formula:
C₄H₁₁N MW: 73.14 g/mol Boiling Pt: 63 °C Melting Pt: –72 °C Density: 0.724 g/mL Flash Pt: –10 °C (2 °F) |
MDL Number:
MFCD00008094 CAS Number: 13952-84-6 EINECS: 237-732-7 UN: 2733 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...