2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%

Supplier: BeanTown Chemical

Synonyms: 1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol

129400-10G 129400-50G
BT129400-10GEA 60 USD
BT129400-10G BT129400-50G
2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%
2,2,3,3,4,4,4-Heptafluoro-1-butanol

CAS: 375-01-9; EC No: 206-782-1; MDL No: MFCD00004674; RTECS: EL4130000 UN No: UN1987; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C4H3F7O; MW: 200.05 Boiling Point: 96-97°; Flash point: 25°C (77°F) Density (g/mL): 1.6; Refractive Index: 1.300


  • RTECS: EL4130000
  • Refractive index: 1.300
  • Liquid

Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 96…97 °C
Density: 1.6 g/mL
Flash Pt: 25 °C (77 °F)
MDL Number: MFCD00004674
CAS Number: 375-01-9
EINECS: 206-782-1
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR