9-(Chloromethyl)anthracene ≥98%

Supplier: BeanTown Chemical

129395-1G 129395-25G 129395-5G
BT129395-1GEA 35 USD
BT129395-1G BT129395-25G BT129395-5G
9-(Chloromethyl)anthracene ≥98%
9-(Chloromethyl)anthracene

CAS: 24463-19-2; EC No: 246-274-7; MDL No: MFCD00001263; RTECS: CA9600000 UN No: UN3261; Haz Class: 8; Packing Group: III Powder; Molecular Formula: C15H11Cl; MW: 226.70 Melting Point: 138-140° Light Sensitive


  • RTECS: CA9600000
  • Light Sensitive
  • Powder

Formula: C₁₅H₁₁Cl
MW: 226.71 g/mol
Melting Pt: 138…140 °C
MDL Number: MFCD00001263
CAS Number: 24463-19-2
EINECS: 246-274-7
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR