Oxalic acid dihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethanedioic acid dihydrate

129385-1KG 129385-250G 129385-5KG
BT129385-1KGEA 77.57 USD
BT129385-1KG BT129385-250G BT129385-5KG
Oxalic acid dihydrate ≥98%
Oxalic acid dihydrate

CAS: 6153-56-6; EC No: 205-634-3; MDL No: MFCD00149102; RTECS: KI1600000 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: HOOCCOOH·2H2O ; Molecular Formula: C2H2O4·2H2O ; MW: 126.07 Melting Point: 104-106° Density (g/mL): 1.65 Hygroscopic


  • RTECS: KI1600000
  • Hygroscopic
  • Crystalline

Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Melting Pt: 104…106 °C
Density: 1.65 g/mL
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR