tert-Butyl carbazate ≥98%
Supplier: BeanTown Chemical
Synonyms:
tert-Butyl hydrazinecarboxylate
129315-100G
129315-25G
129315-500G
BT129315-100GEA
137.72
USD
BT129315-100G
BT129315-25G
BT129315-500G
tert-Butyl carbazate ≥98%
tert-Butyl carbazate
CAS: 870-46-2; EC No: 212-795-3; MDL No: MFCD00007593
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Crystalline; Linear Formula: (CH3)3COCONHNH2; Molecular Formula: C5H12N2O2; MW: 132.16
Melting Point: 37-41°; Boiling Point: 63-65°/0.1 mmHg; Flash point: 91°C (195°F)
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
C₅H₁₂N₂O₂ Boiling Pt: 63…65 °C/0.1 mmHg Melting Pt: 37…41 °C Flash Pt: 91 °C (195 °F) |
MDL Number:
MFCD00007593 CAS Number: 870-46-2 EINECS: 212-795-3 UN: 1325 ADR: 4.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...