(±)-Camphor-10-sulfonic acid ≥98%

Supplier: BeanTown Chemical

129295-100G 129295-500G
BT129295-100GEA 39 USD
BT129295-100G BT129295-500G
(±)-Camphor-10-sulfonic acid ≥98%
(±)-Camphor-10-sulphonic acid

CAS: 5872-08-2; EC No: 227-527-0; MDL No: MFCD00074827; RTECS: DT5077100 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Molecular Formula: C10H16O4S; MW: 232.30 Melting Point: 203-206° (decomposes) Hygroscopic


  • RTECS: DT5077100
  • Hygroscopic
  • Crystalline

Formula: C₁₀H₁₆O₄S
Melting Pt: 203…206 °C (dec.)
MDL Number: MFCD00074827
CAS Number: 5872-08-2
EINECS: 227-527-0
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR