Iodine monochloride ACS
Supplier: BeanTown Chemical
Synonyms:
Iodochloride
129290-100G
129290-25G
129290-500G
BT129290-100GEA
139
USD
BT129290-100G
BT129290-25G
BT129290-500G
Iodine monochloride ACS
Iodine monochloride
CAS: 7790-99-0; EC No: 232-236-7; MDL No: MFCD00011354; RTECS: NN1650000
UN No: UN3498; Haz Class: 8; Packing Group: II
Crystalline/Liquid; Linear Formula: ICl; MW: 162.36
Boiling Point: 96-98°
Density (g/mL): 3.24
Light Sensitive, Moisture Sensitive
- RTECS: NN1650000
- Light Sensitive, Moisture Sensitive
- Crystalline/Liquid
Formula:
ClI MW: 162.36 g/mol Boiling Pt: 96…98 °C Density: 3.24 g/mL |
MDL Number:
MFCD00011354 CAS Number: 7790-99-0 EINECS: 232-236-7 UN: 3498 ADR: 8,II |
Specification Test Results
I/Cl ratio | 1.0-1.2 |
Insoluble matter | ≤0.005% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...