Sodium azide ≥99%
Supplier: BeanTown Chemical
CAS: 26628-22-8; EC No: 247-852-1; MDL No: MFCD00003536; RTECS: VY8050000
UN No: UN1687; Haz Class: 6.1; Packing Group: II
Crystalline/Powder; Linear Formula: NaN3; MW: 65.01
Melting Point: 275° (decomposes)
Density (g/mL): 1.85
Heat Sensitive, Moisture Sensitive
- RTECS: VY8050000
- Heat Sensitive, Moisture Sensitive
- Crystalline/Powder
Formula:
NaN₃ Melting Pt: 275 °C (dec.) Density: 1.85 g/mL |
MDL Number:
MFCD00003536 CAS Number: 26628-22-8 EINECS: 247-852-1 UN: 1687 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...