Cerium(IV)-2-methoxyethoxide 18 - 20% (w/w) in 2-methoxyethanol
Supplier: BeanTown Chemical
129185-25G
129185-5G
BT129185-25GEA
379
USD
BT129185-25G
BT129185-5G
Cerium(IV)-2-methoxyethoxide 18 - 20% (w/w) in 2-methoxyethanol
Cerium(IV)-2-methoxyethoxide
CAS: 876107-33-4; MDL No: MFCD00236530
UN No: UN1188; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C12H28CeO8 ; MW: 440.47
Density (g/mL): 1.02
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₁₂H₂₈CeO₈ Density: 1.02 g/mL |
MDL Number:
MFCD00236530 CAS Number: 876107-33-4 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...