2,3-Diaminopyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: Pyridine-2,3-diyldiamine

129085-100G 129085-25G 129085-5G
BT129085-100GEA 600 USD
BT129085-100G BT129085-25G BT129085-5G
2,3-Diaminopyridine ≥98%
2,3-Diaminopyridine

CAS: 452-58-4; EC No: 207-200-9; MDL No: MFCD00006319; RTECS: US7548000 UN No: UN2671; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: C5H7N3; MW: 109.13 Melting Point: 110-115° Air Sensitive


  • RTECS: US7548000
  • Air Sensitive
  • Powder

Formula: C₅H₇N₃
MW: 109.13 g/mol
Melting Pt: 110…115 °C
MDL Number: MFCD00006319
CAS Number: 452-58-4
EINECS: 207-200-9
UN: 2671
ADR: 6.1 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR