2-Bromopropene ≥99% stabilized

Supplier: BeanTown Chemical

Synonyms: 2-Propenyl bromide, α-Methylvinyl bromide, Isopropenyl bromide

129075-100G 129075-25G 129075-5G
BT129075-100GEA 482 USD
BT129075-100G BT129075-25G BT129075-5G
2-Bromopropene ≥99% stabilized
2-Bromopropene

CAS: 557-93-7; EC No: 209-185-4; MDL No: MFCD00000140; RTECS: UC7085000 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3C(Br)=CH2; Molecular Formula: C3H5Br; MW: 120.98 Boiling Point: 47-49°; Flash point: 4°C (39°F) Density (g/mL): 1.362; Refractive Index: 1.444 Light Sensitive


  • RTECS: UC7085000
  • Refractive index: 1.444
  • Light Sensitive
  • Liquid

Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 47…49 °C
Density: 1.362 g/mL
Flash Pt: 4 °C (39 °F)
MDL Number: MFCD00000140
CAS Number: 557-93-7
EINECS: 209-185-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR