Iron(III) perchlorate hydrate
Supplier: BeanTown Chemical
129050-100G
129050-500G
BT129050-100GEA
52
USD
BT129050-100G
BT129050-500G
Iron(III) perchlorate hydrate
Iron(III) perchlorate hydrate
CAS: 15201-61-3; EC No: 236-908-0; MDL No: MFCD00011000
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: Fe(ClO4)3·xH2O; MW: 354.19 (anhydrous)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Cl₃FeH₂O₁₃ |
MDL Number:
MFCD00149705 CAS Number: 15201-61-3 EINECS: 236-908-0 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...