1-(2-Fluorophenyl)piperazine ≥99%
Supplier: BeanTown Chemical
129010-25G
129010-5G
BT129010-25GEA
150
USD
BT129010-25G
BT129010-5G
1-(2-Fluorophenyl)piperazine ≥99%
1-(2-Fluorophenyl)piperazine
CAS: 1011-15-0; EC No: 213-780-4; MDL No: MFCD00040726
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III
Crystalline; Molecular Formula: C10H13FN2; MW: 180.22
Boiling Point: 150°/3 mmHg; Flash point:
- Refractive index: 1.566
- Air Sensitive
- Crystalline
Formula:
C₁₀H₁₃FN₂ MW: 180.23 g/mol Boiling Pt: 150 °C/3 mmHg Density: 1.141 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00040726 CAS Number: 1011-15-0 EINECS: 213-780-4 UN: 2923 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...