3,4-Dihydroxybenzonitrile ≥97%
Supplier: BeanTown Chemical
128955-1G
128955-5G
BT128955-1GEA
23
USD
BT128955-1G
BT128955-5G
3,4-Dihydroxybenzonitrile ≥97%
3,4-Dihydroxybenzonitrile
CAS: 17345-61-8; EC No: 241-367-9; MDL No: MFCD00016436
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C7H5NO2; MW: 135.12
Melting Point: 155-159°
- Powder
Formula:
C₇H₅NO₂ Melting Pt: 155…159 °C |
MDL Number:
MFCD00016436 CAS Number: 17345-61-8 EINECS: 241-367-9 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...