1,3-Dibromo-5,5-dimethylhydantoin ≥98%
Supplier: BeanTown Chemical
CAS: 77-48-5; EC No: 201-030-9; MDL No: MFCD00003189; RTECS: MU0686000
UN No: UN3085; Haz Class: 5.1 (8); Packing Group: II
Powder; Molecular Formula: C5H6Br2N2O2; MW: 285.92
Melting Point: 197-199° (decomposes)
Moisture Sensitive
- RTECS: MU0686000
- Moisture Sensitive
- Powder
Formula:
C₅H₆Br₂N₂O₂ Melting Pt: 197…199 °C (dec.) |
MDL Number:
MFCD00003189 CAS Number: 77-48-5 EINECS: 201-030-9 ADR: 5.1 (8),II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...