3,3-Dimethylbutanone ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,1,1-Trimethylacetone, Methyl tert-butyl ketone, Pinacolone, tert-Butyl methyl ketone, 3,3-Dimethyl-2-butanone

128905-100G 128905-500G
BT128905-100GEA 113.3 USD
BT128905-100G BT128905-500G
3,3-Dimethylbutanone ≥97%
3,3-Dimethylbutanone

CAS: 75-97-8; EC No: 200-920-4; MDL No: MFCD00008846 UN No: UN1224; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3COC(CH3)3; Molecular Formula: C6H12O; MW: 100.16 Melting Point: -53°; Boiling Point: 103-107°; Flash point: 5°C (41°F) Density (g/mL): 0.801; Refractive Index: 1.396


  • Refractive index: 1.396
  • Liquid

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 103…107 °C
Melting Pt: –53 °C
Density: 0.801 g/mL
Flash Pt: 5 °C (41 °F)
MDL Number: MFCD00008846
CAS Number: 75-97-8
EINECS: 200-920-4
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR