(±)-trans-1,2-Diaminocyclohexane ≥99%
Supplier: BeanTown Chemical
Synonyms:
(±)-trans-1,2-Cyclohexanediamine, trans-Cyclohexane-1,2-diamine
128895-10G
128895-250G
128895-50G
BT128895-10GEA
27.28
USD
BT128895-10G
BT128895-250G
BT128895-50G
(±)-trans-1,2-Diaminocyclohexane ≥99%
(±)-trans-1,2-Diaminocyclohexane
CAS: 1121-22-8; MDL No: MFCD00063747
UN No: UN2735; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C6H14N2; MW: 114.19
Melting Point: 14-15°; Boiling Point: 79-81°/15 mmHg; Flash point: 69°C (156°F)
Density (g/mL): 0.951; Refractive Index: 1.489
Air Sensitive
- Refractive index: 1.489
- Air Sensitive
- Liquid
Formula:
C₆H₁₄N₂ MW: 114.19 g/mol Boiling Pt: 79…81 °C/15 mmHg Melting Pt: 14…15 °C Density: 0.951 g/mL Flash Pt: 69 °C (156 °F) |
MDL Number:
MFCD00063747 CAS Number: 1121-22-8 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...