Cadmium(II) chloride, anhydrous ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Cadmium chloride
128855-100G
128855-25G
BT128855-100GEA
426
USD
BT128855-100G
BT128855-25G
Cadmium(II) chloride, anhydrous ≥99.99% (trace metals basis)
Cadmium(II) chloride
CAS: 10108-64-2; EC No: 233-296-7; MDL No: MFCD00010916; RTECS: EV0175000
UN No: UN2570; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: CdCl2; MW: 183.32
Melting Point: 568°
Hygroscopic
- RTECS: EV0175000
- Hygroscopic
- Powder
Formula:
CdCl₂ MW: 183.32 g/mol Melting Pt: 568 °C |
MDL Number:
MFCD00010916 CAS Number: 10108-64-2 EINECS: 233-296-7 UN: 2570 ADR: 6.1,III |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...