Cyclopentadienylmanganese tricarbonyl

Supplier: BeanTown Chemical

Synonyms: Cymantrene, Cyclopentadienylmanganese(I) tricarbonyl

128770-1G 128770-25G 128770-5G
BT128770-1GEA 46 USD
BT128770-1G BT128770-25G BT128770-5G
Cyclopentadienylmanganese tricarbonyl
Cyclopentadienylmanganese tricarbonyl

CAS: 12079-65-1; EC No: 235-142-4; MDL No: MFCD00001440 UN No: UN3466; Haz Class: 6.1; Packing Group: II Crystalline; Molecular Formula: C8H5MnO3; MW: 204.06 Melting Point: 72-76° Air Sensitive


  • Air Sensitive
  • Crystalline

Formula: C₈H₅MnO₃
MW: 204.06 g/mol
Melting Pt: 72…76 °C
MDL Number: MFCD00001440
CAS Number: 12079-65-1
EINECS: 235-142-4
UN: 3466
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR