o-Terphenyl ≥98%
Supplier: BeanTown Chemical
CAS: 84-15-1; EC No: 201-517-6; MDL No: MFCD00003055; RTECS: WZ6472000
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Linear Formula: C6H5C6H4C6H5; Molecular Formula: C18H14; MW: 230.31
Melting Point: 56-59°; Boiling Point: 337°; Flash point:
- RTECS: WZ6472000
- Crystalline
Formula:
C₁₈H₁₄ Boiling Pt: 337 °C Melting Pt: 56…59 °C Density: 1.1 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00003055 CAS Number: 84-15-1 EINECS: 201-517-6 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...