2-(Trimethylsilyl)ethanol ≥98%
Supplier: BeanTown Chemical
128740-100G
128740-25G
128740-5G
BT128740-100GEA
835
USD
BT128740-100G
BT128740-25G
BT128740-5G
2-(Trimethylsilyl)ethanol ≥98%
2-(Trimethylsilyl)ethanol
CAS: 2916-68-9; EC No: 220-844-5; MDL No: MFCD00002825; RTECS: KM5480000
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: (CH3)3SiCH2CH2OH; Molecular Formula: C5H14OSi; MW: 118.25
Boiling Point: 71-73°/35 mmHg; Flash point: 56°C (133°F)
Density (g/mL): 0.825; Refractive Index: 1.423
- RTECS: KM5480000
- Refractive index: 1.423
- Liquid
Formula:
HOH₂CCH₂Si(CH₃)₃ MW: 118.25 g/mol Boiling Pt: 71…73 °C/35 mmHg Melting Pt: 0 Density: 0.825 g/mL Flash Pt: 56 °C (133 °F) |
MDL Number:
MFCD00002825 CAS Number: 2916-68-9 EINECS: 220-844-5 UN: 1987 ADR: 3 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...