1-(2-Methoxyphenyl)piperazine ≥98%

Supplier: BeanTown Chemical

128725-100G 128725-25G 128725-5G
BT128725-100GEA 385 USD
BT128725-100G BT128725-25G BT128725-5G
1-(2-Methoxyphenyl)piperazine ≥98%
1-(2-Methoxyphenyl)piperazine

CAS: 35386-24-4; EC No: 252-537-7; MDL No: MFCD00005958 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III Solid; Molecular Formula: C11H16N2O; MW: 192.26 Melting Point: 35-40°; Boiling Point: 130-133°/0.1 mmHg; Flash point: 112°C (234°F) Density (g/mL): 1.095; Refractive Index: 1.575 Air Sensitive


  • Refractive index: 1.575
  • Air Sensitive
  • Solid

Formula: C₁₁H₁₆N₂O
Boiling Pt: 130…133 °C/0.1 mmHg
Melting Pt: 35…40 °C
Density: 1.095 g/mL
Flash Pt: 112 °C (234 °F)
MDL Number: MFCD00005958
CAS Number: 35386-24-4
EINECS: 252-537-7
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR